Learn More
Thermo Scientific™ Shandon™ ColorFrost™ Plus Slides Available on GSA/VA Contract for Federal Government customers only.
Improve cellular adhesion with positively charged Thermo Scientific™ Shandon™ ColorFrost™ Plus Slides.
Manufacturer:  Thermo Scientific™ 9991009
 View more versions of this product
Catalog No. 99-910-09
Add to cart
Description
Description
Improve cellular adhesion with Thermo Scientific™ Shandon™ ColorFrost™ Plus Slides, made of positively charged uncoated glass.
  • Electrostatically attract tissue sections and cytology preparations, binding them to the slide
  • Eliminate the need for special adhesives or protein coatings
  • No background staining
  • Available in a variety of colors
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Lavender
100/Cs.