Learn More
Thermo Scientific™ Shandon™ Curved Mayo Dissecting Scissors
Available in premium and standard grades. Cut precisely with Thermo Scientific™ Shandon™ Curved Mayo Dissecting Scissors.
Manufacturer:  Thermo Scientific™ 787030
 View more versions of this product
Catalog No. 787030
  Check Availability
Add to cart
Specifications
Description
Available in premium and standard grades. Cut precisely with Thermo Scientific™ Shandon™ Curved Mayo Dissecting Scissors.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Guaranteed to be free from defects in material and workmanship
Description & Specifications
Specifications
Curved
9 in.
Curved Mayo Dissecting Scissors
Premium
22.9cm