Learn More
Epredia™ Cytospin™ Collection Fluid
Store specimens or unstained slides for weeks without adverse effect with Epredia™ Shandon™ Cytospin™ Collection Fluid.
Supplier:  Epredia™ 9990323
 View more versions of this product
Catalog No. 9990323
Add to Cart
Description
Description
  • Packaged in convenient pump bottle or pre-filled specimen cups
  • Ready-to-use alcohol/Carbowax™ (PEG) combination for direct specimen collection and fixation
Specifications
Specifications
Collection Fluid
120mL
Alcohol/Carbowax (PEG) combination
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov