Learn More
Thermo Scientific™ Shandon™ Disposable Base Molds
Promote rapid paraffin hardening with Thermo Scientific™ Shandon™ Disposable Base Molds.
Manufacturer:  Thermo Scientific™ 41744
 View more versions of this product
Catalog No. 41-744
  Check Availability
Add to cart
Specifications
Description
Promote rapid paraffin hardening with Thermo Scientific™ Shandon™ Disposable Base Molds.
  • Constructed of rigid transparent plastic
Description & Specifications
Specifications
Plastic
0.95 in.
1.46 in.
0.2 in.
Base Mold, Disposable
24mm
37mm
5mm