Thermo Scientific™ Shandon™ Dissecting Tray for Raised Dissecting Boards
Ample space allows for instruments and drainage on Shandon™ Dissecting Tray for Raised Dissecting Boards.
Manufacturer: Thermo Scientific™ 355
 View more versions of this product
Catalog No. 35-5
  Check Availability
Add to Cart
    Specifications
    Length (English) 16.25 in.
    Width (English) 13.25 in.
    Item Description Dissecting Tray
    Ample space allows for instruments and drainage on Thermo Scientific™ Shandon™ Dissecting Tray for Raised Dissecting Boards.
    Features:
    • Polyethylene tray comes in small and large sizes
    • Four embossments on tray bottom hold Raised Dissecting Boards 356 and 35614
  • Description & Specifications
Related Products
Product Content Feedback