Thermo Scientific™ Shandon™ Double Curved Post-Mortem Needle
Stitch with Thermo Scientific™ Shandon™ Double Curved Post-Mortem Needles for quality and precision.
Manufacturer: Thermo Scientific™ 4113
 View more versions of this product
Catalog No. 41-13
  Check Availability
Add to Cart
    Specifications
    Length (English) 4 in.
    Length (Metric) 10.2cm
    Tip Style Double Curved
    Type Postmortem,Postmortem
    Stitch with Thermo Scientific™ Shandon™ Double Curved Post-Mortem Needles for quality and precision. Needles are available in five lengths.
    Features:
    • Properly hardened, tempered and passivated
    • Electronically polished and cleaned for a high-quality finish
    • Patterns, dimensions and finish held to exacting standards
    • Guaranteed to be free from defects in material and workmanship
  • Description & Specifications
Related Products
Product Content Feedback