Learn More
Thermo Scientific™ Shandon™ Double Cytofunnel™
Optimize cell yield with the Thermo Scientific™ Shandon™ Double Cytofunnel™.
$172.30
Specifications
Type Disposable Sample Chamber, With Filter Cards and Caps
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
59-910-39
View Documents
Thermo Scientific™
5991039
25/Box
Case of 25 for $172.30
Add to cart
Description
Description
Optimize cell yield with the Thermo Scientific™ Shandon™ Double Cytofunnel™. Features disposable sample chambers with white filter cards and caps.
  • Produces more cost-effective, thin-layer preparations when compared to other thin-layer methods
  • Reduces the risk of exposure to pathogenic samples
  • Reduces preparation time
  • Disposable to eliminate time-consuming decontamination and cleaning
  • Shandon™ Double Cytofunnel™ REQUIRES Thermo Scientific™ Shandon™ Cytoclips™
  • Decreases the possibility of cross-contamination
  • Features funnel markings corresponding to slide markings for simple identification
Includes:
  • Pre-attached filter card
  • Closure caps
  • Easy-to-use operational illustrations
Specifications
Specifications
Disposable Sample Chamber, With Filter Cards and Caps