Learn More
Thermo Scientific™ Shandon™ Double Cytoslides™ Available on GSA/VA Contract for Federal Government customers only.
Use Thermo Scientific™ Shandon™ Double Cytoslides™ with Thermo Scientific™ Shandon™ EZ Double Cytofunnel™ Chambers.
$90.10
Specifications
Color Plain
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
59-910-55 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
5991055
Coated
Case of 100 for $90.10
Add to cart
Description
Description
Use Thermo Scientific™ Shandon™ Double Cytoslides™ with Thermo Scientific™ Shandon™ EZ Double Cytofunnel™ Chambers.
  • Specimen is displayed within a circle marked as 1 or 2
  • Number circles correspond with the Cytofunnel™ chamber
Ordering Alert:
  • Sold in boxes of 100 slides
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Plain