Learn More
Thermo Scientific™ Shandon™ Embedding Cassettes II in Tube Packs
Hold tissue specimens during processing and embedding with Thermo Scientific™ Shandon™ Embedding Cassettes II in Tube Packs.
Manufacturer:  Thermo Scientific™ B1000729YW
 View more versions of this product
Catalog No. B1000729YW
Add to cart
Description
Description
Hold tissue specimens during processing and embedding with Thermo Scientific™ Shandon™ Embedding Cassettes II in Tube Packs. The cassettes are compatible with Thermo Scientific™ Shandon™ MicroWriter™ cassette labelers.
  • Acetal polymer cassettes with writing face at 45° angle
  • Slotted pore style
  • Locking lid attached by two hinges
Ordering Information:
  • Tube packs are supplied in 10 tubes of 75 cassettes each
Specifications
Specifications
Slotted
45°
Acetal polymer
Biopsy Cassettes, Slotted Pore
Yellow