Thermo Scientific™ Shandon™ Enterotomy No Hook Scissors
For precise incisions, use Thermo Scientific™ Shandon™ Enterotomy Scissors, available in premium and standard grades.
Manufacturer: Thermo Scientific™ 71901
 View more versions of this product
Catalog No. 22-045-455
  Check Availability
Add to Cart
    Specifications
    Length (English) 8 in.
    Length (Metric) 20.3cm
    Blade Style No Hook
    Grade Premium
    For precise incisions, use Thermo Scientific™ Shandon™ Enterotomy Scissors, available in premium and standard grades.
    • Properly hardened, tempered and passivated
    • Electronically polished and cleaned for a high-quality finish
    • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
    • Guaranteed to be free from defects in material and workmanship
  • Description & Specifications
Related Products
Product Content Feedback