Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Shandon™ EZ Single Cytofunnel™ with White Filter Cards and Caps Available on GSA/VA Contract for Federal Government customers only.

Optimize cell yield with the Thermo Scientific™ Shandon™ EZ Single Cytofunnel™ with White Filter Cards and Caps.

Manufacturer:  Thermo Scientific™ A78710003

 View more versions of this product

Catalog No. 14-390-45


Add to cart

Description

Description

Optimize cell yield with the Thermo Scientific™ Shandon™ EZ Single Cytofunnel™ with White Filter Cards and Caps. A disposable sample chamber with cap and pre-attached white filter card for high absorbency, this product is meant for single use with all Thermo Scientific™ Shandon™ Cytospin™ Cytocentrifuge applications.

  • Reduces the risk of exposure to pathogenic samples
  • Disposable to eliminate time-consuming decontamination and cleaning
  • Decreases the possibility of cross-contamination
  • Eliminates the need for metal Thermo Scientific™ Shandon™ Cytoclips™
  • Produces more cost-effective, thin-layer preparations when compared to other thin-layer methods
  • Holds sample volumes up to 0.5mL
  • Includes cell deposition area of 6mm in diameter
Includes:
  • Closure caps
  • Easy-to-use operational illustrations

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

40/box
40/Box
Disposable|With Filter Cards and Caps
White
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit