Learn More
Epredia™ EZ Single Cytofunnel™ with White Filter Cards and Caps
Optimize cell yield with the Epredia™ EZ Single Cytofunnel™ with White Filter Cards and Caps.
$234.00 - $658.00
Specifications
Volume (Metric) 0.1 to 0.5 mL
Color White
Type EZ Single Cytofunnel™, With Filter Cards and Caps
Disposable Yes
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
14-390-45
View Documents
Epredia™
A78710003
40 Pk.
Case of 40 for $234.00
Add to cart
A78710020 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Epredia™
A78710020
200/Cs.
Case of 200 for $658.00
Add to cart
Description
Description
  • Reduces the risk of exposure to pathogenic samples
  • Disposable to eliminate time-consuming decontamination and cleaning
  • Decreases the possibility of cross-contamination
  • Eliminates the need for metal Cytoclips™
  • Produces more cost-effective, thin-layer preparations when compared to other thin-layer methods
  • Holds sample volumes up to 0.5 mL
  • Includes cell deposition area of 6 mm in diameter
Includes:
  • Closure caps
  • Easy-to-use operational illustrations
Specifications
Specifications
0.1 to 0.5 mL
White
EZ Single Cytofunnel™, With Filter Cards and Caps
Yes