Learn More
Thermo Scientific™ Shandon™ Finesse™ ME/E Series Disposable Blade Holders Available on GSA/VA Contract for Federal Government customers only.
Eliminate the need to switch to a different knife holder for larger specimens by using the Thermo Scientific™ Shandon™ Finesse™ ME/E Series Disposable Blade Holders.
Manufacturer:  Thermo Scientific™ A77510272
 View more versions of this product
Catalog No. A77510272
Add to cart
Description
Description
Eliminate the need to switch to a different knife holder for larger specimens by using the Thermo Scientific™ Shandon™ Finesse™ ME/E Series Disposable Blade Holders. Use these holders with standard cassettes or Thermo Scientific™ Super Mega™ cassettes. Holders can be used with either the Thermo Scientific™ Shandon™ Finesse™ ME+/ME or the Thermo Scientific™ Shandon™ Finesse™ E+ E microtome.
  • Improved clamping mechanism design significantly reduces need for knife holder maintenance and improves reliability
  • Consistent reproducible sections provided
  • Larger clamp plate allows easier section retrieval
  • Clamped blade is fully supported along its entire length
  • Entire top stage moves laterally for safer operation
  • Minimized wear on clamping mechanism
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Low-profile
Standard or Thermo Scientific™ cassettes
Low-profile
Low profile