Learn More
Thermo Scientific™ Shandon™ Finesse™ ME/E Series Disposable Blade Holders
Eliminate the need to switch to a different knife holder for larger specimens by using the Thermo Scientific™ Shandon™ Finesse™ ME/E Series Disposable Blade Holders.
Manufacturer:  Thermo Scientific™ A77510275
 View more versions of this product
Catalog No. A77510275
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Eliminate the need to switch to a different knife holder for larger specimens by using the Thermo Scientific™ Shandon™ Finesse™ ME/E Series Disposable Blade Holders. Use these holders with standard cassettes or Thermo Scientific™ Super Mega™ cassettes. Holders can be used with either the Thermo Scientific™ Shandon™ Finesse™ ME+/ME or the Thermo Scientific™ Shandon™ Finesse™ E+ E microtome.
  • Improved clamping mechanism design significantly reduces need for knife holder maintenance and improves reliability
  • Consistent reproducible sections provided
  • Larger clamp plate allows easier section retrieval
  • Clamped blade is fully supported along its entire length
  • Entire top stage moves laterally for safer operation
  • Minimized wear on clamping mechanism
Specifications
Specifications
High-profile
Standard or Thermo Scientific™ cassettes
High-profile
High profile