Learn More
Thermo Scientific™ Shandon™ Formal-Fixx™ 10% Neutral Buffered Formalin
Produce consistent results and extend life of rotary valve in tissue processors with Thermo Scientific™ Shandon™ Formal-Fixx™ 10% Neutral Buffered Foramalin.
Manufacturer:  Thermo Scientific™ 9990234
 View more versions of this product
Catalog No. 9990234
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Produce consistent results and extend life of rotary valve in tissue processors with Thermo Scientific™ Shandon™ Formal-Fixx™ 10% Neutral Buffered Foramalin.
  • Phosphate-free
  • Use for storage of all tissue types
  • Stores specimens without excessive shrinking or hardening
  • Available premixed, ready-to-use, or in economical concentrate
  • Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
  • Pre-filled, leak-resistant cups are available
Specifications
Specifications
Neutral Buffered Formalin
Ready-to-Use
2.6 gal.