Learn More
Thermo Scientific™ Shandon™ Glass Storage Jars
Store and display specimens in Thermo Scientific™ Shandon™ Glass Storage Jars.
Manufacturer:  Thermo Scientific™ 885
 View more versions of this product
Catalog No. 88-5
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Store and display specimens in Thermo Scientific™ Shandon™ Glass Storage Jars.
  • Clear glass jar equipped with a captive gasket
  • Cover held in place by a hinged spring clamp
Description & Specifications
Specifications
16 oz., 474mL
16 oz.
Glass Storage Jars
474mL
Glass