Learn More
Epredia™ Glyo-Fixx™
Obtain superior results to neutral buffered formalin with Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
Supplier:  Epredia™ 9990920
 View more versions of this product
Catalog No. 9990920
Add to Cart
Description
Description
  • Produces distinct and crisp cellular morphology
  • Fixes most antigens without impairing immunoreactivity
  • Allows special stain dyes, including silver procedures, to be sharp without background staining
  • Presents fine detail in specimen structure
Specifications
Specifications
Fixative
180 x 20mL