Learn More
Thermo Scientific™ Shandon™ Gram Stain Available on GSA/VA Contract for Federal Government customers only.
Differentiate between Gram-positive and Gram-negative bacteria with Thermo Scientific™ Shandon™ Gram Stain.
Manufacturer:  Thermo Scientific™ 9990726
 View more versions of this product
Catalog No. 9990726
  Check Availability
Add to cart
Specifications
Description
Differentiate between Gram-positive and Gram-negative bacteria with Thermo Scientific™ Shandon™ Gram Stain, which may be used as a routine bacteriologic stain on fixed smears and histological slides.
  • Safranin O provided as a counterstain
  • Based upon the popular Hucker modification
  • Demonstrates the presence of microorganisms such as bacteria
Ordering Information:
Available in kit form or as individual components.
Includes:
Crystal Violet Stain; Gram's Iodine (stabilized); Decolorizing Solution; Safranin O Counterstain
Description & Specifications
Specifications
Gram's iodine
4L
Crystal violet iodine