Learn More
Thermo Scientific™ Shandon™ Gram Stain
Differentiate between Gram-positive and Gram-negative bacteria with Thermo Scientific™ Shandon™ Gram Stain.
$164.00
Products 1
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
9990725
View Documents
Thermo Scientific™
9990725
Gram crystal violet stain 4L Each for $164.00
Add to cart
Description
Description
Differentiate between Gram-positive and Gram-negative bacteria with Thermo Scientific™ Shandon™ Gram Stain, which may be used as a routine bacteriologic stain on fixed smears and histological slides.
  • Safranin O provided as a counterstain
  • Based upon the popular Hucker modification
  • Demonstrates the presence of microorganisms such as bacteria
Ordering Information:
Available in kit form or as individual components.
Includes:
Crystal Violet Stain; Gram's Iodine (stabilized); Decolorizing Solution; Safranin O Counterstain