Thermo Scientific™ Shandon™ Halstead's Hemostatic Mosquito Straight Forceps

Choose 5 in. (12.7cm) curved Halstead's Hemostatic Mosquito Straight Forceps for quality and precision.

Manufacturer: Thermo Scientific™ 7507

 View more versions of this product

Catalog No. 7507

  Check Availability
Add to Cart

    Specifications

    Grade Premium
    Length (English) 5 in.
    Length (Metric) 12.7cm
    Shape Straight
    Type Halstead

    Choose 5 in. (12.7cm) curved Thermo Scientific™ Shandon™ Halstead's Hemostatic Mosquito Straight Forceps for quality and precision. Forceps are available in premium and standard grades.

    • Properly hardened, tempered and passivated
    • Electronically polished and cleaned for a high-quality finish
    • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
    • Guaranteed to be free from defects in material and workmanship

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit