Learn More
Epredia™ Accessories for Hyperclean TruAir™ Workstation
For use with Epredia™ Hyperclean Pathology Workstations.
Supplier: Epredia™ 9990648
Description
Accessories:- Charcoal Filter
- Fine particle Filters
- Formaldehyde Neutralizing (Potassium Permanganate) Filter
Specifications
Workstation Accessories | |
Replacement Hood; Hyperclean 1 | |
Epredia™ Hyperclean 1 Fume Control Workstation | |
Hyperclean™ |