Learn More
Thermo Scientific™ Shandon™ Hyperclean™ Workstation Accessories
Use Thermo Scientific™ Shandon™ Hyperclean™ Workstation Accessories with Shandon™ Hyperclean Pathology Workstations.
Manufacturer:  Thermo Scientific™ 9990611
 View more versions of this product
Catalog No. 99-906-11
  Check Availability
Add to cart
Specifications
Description
Use these Thermo Scientific™ Shandon™ Hyperclean™ Workstation Accessories with Shandon™ Hyperclean™ Pathology Workstations.
Accessories:
  • Charcoal Filter
  • Fine particle Filters
  • Formaldehyde Neutralizing (Potassium Permanganate) Filter
  • Vent Adapter Kit
  • Drip Tray
  • Stainless steel insert
  • Replacement Hood
  • Safety Door with Hand Holes
Description & Specifications
Specifications
Hyperclean™
Fine particle filters
Fine particle filters
Workstation Accessories
Shandon Hyperclean Pathology Workstations