Learn More
Thermo Scientific™ Shandon™ Instant Eosin Available on GSA/VA Contract for Federal Government customers only.
Obtain brilliant and consistent staining with Thermo Scientific™ Shandon™ Instant Eosin.
Manufacturer:  Thermo Scientific™ 6765540
 View more versions of this product
Catalog No. 6765540
  Check Availability
Add to cart
Specifications
Description
Obtain brilliant and consistent staining with Thermo Scientific™ Shandon™ Instant Eosin.
  • Stable and precipitate-free; no filtering
  • pH-adjusted for maximum stain uptake
  • Economical powder stores indefinitely
  • Compatible with all alum Hematoxylin stains
Description & Specifications
Specifications
Eosin
Alum Hematoxylin Stains
Liquid
Aqueous