Learn More
Thermo Scientific™ Shandon™ Large Section Blades
Remove skull cap or bone specimens with general purpose Thermo Scientific™ Shandon™ Large Section Blades.
Manufacturer:  Thermo Scientific™ 1002100
 View more versions of this product
Catalog No. 10-021-00
  Check Availability
Add to cart
Specifications
Description
Remove skull cap or bone specimens with general purpose Thermo Scientific™ Shandon™ Large Section Blades.
  • Bottom blade width: 2.5in. (6.3cm)
  • Top blade width: 0.43in. (1.1cm)
  • Permanently bonded stainless-steel arbor (10-003 only)
Description & Specifications
Specifications
No
0.43 in.
1.1cm
2.5 in.
6.3cm