Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Thermo Scientific™ Shandon™ Microtome Knives
Assure performance through multiple tests with Thermo Scientific™ Shandon™ Microtome Knives.
Manufacturer:  Thermo Scientific™ 9992009
 View more versions of this product
Catalog No. 9992009
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Assure performance through multiple tests with Thermo Scientific™ Shandon™ Microtome Knives.
  • Precision ground for outstanding sectioning and extended cutting life; flat-back design
  • C-profile and D-profile knives are all drilled and tapped in the base for direct attachment to holders for sharpening
Description & Specifications
Specifications
C
C-Profile
12cm