Learn More
Thermo Scientific™ Shandon™ Microtome Lubricating Oil
Choose Thermo Scientific™ Shandon™ Microtome Lubricating Oil for use on all Thermo Scientific™ Shandon™ microtomes unless otherwise indicated.
Manufacturer:  Thermo Scientific™ 288
Catalog No. 28-8
Add to cart
Description
Description
Choose Thermo Scientific™ Shandon™ Microtome Lubricating Oil for use on all Thermo Scientific™ Shandon™ microtomes unless otherwise indicated.
  • Not for use with Thermo Scientific Shandon Finesse™ ME/E Series microtomes
  • Not for use with cryostats
Specifications
Specifications
Lubricating oil