Thermo Scientific™ Shandon™ Mobile Carts
Use Thermo Scientific™ Shandon™ Mobile Carts with AN-49 Transportable Autopsy Tables.
Manufacturer: Thermo Scientific™ 11057
 View more versions of this product
Catalog No. 23-245-394
  Check Availability
Add to Cart
    Specifications
    Description Standard AN-49-C Mobile Cart
    For Use With (Equipment) AN-49 Transportable Autopsy Tables
    For Use With AN-49 Transportable Autopsy Tables
    Item Description Standard Mobile Cart
    Use Thermo Scientific™ Shandon™ Mobile Carts with AN-49 Transportable Autopsy Tables.
    Speed and Versatility
    • Intuitive software can be easily downloaded for manual use or remote control through TCP/IP connection
    • Scans a single 2D barcoded storage tube or entire tube rack in less than one second
    • Robust and compact integration-friendly design are ideal for high throuput use
    Enhanced Tube Readability
    • Configured for all ThermoScientific 2D barcoded tubes and user configurable for other 2D formats
    • Integration of linear 1D barcode reader allows integration of individual tube and rack barcode data
    Simple Operation
    • Connect to your computer via a single USB port and, despite which way the tube rack is placed, the computer automatically adjusts orientation and correctly aligns codes with well positions
    • Connects to PC via a single USB port of easy installation
    • Single camera enables save of image file
    • Automatic rack orientation detection and adjustment of 2D barcode read data
    Superior Sample Protection
    • Ability to operate down to -40°C enables barcode reading in cold rooms and low temperature liquid stores to preserve sample integrity
    • Sealed unit prevents frost and condensation which allows reading of frozen tubes at ambient temperatures
    Data File Flexibility
    • Easily integrated into LIMS, databases, or tracking systems
    • User-defined code types and lengths indicate incorrect codes and limit access by user profile
    • Optimized software allows users the ability to select the export data format and conserve memory space
  • Description & Specifications
Related Products
Product Content Feedback