Learn More
Thermo Scientific™ Shandon™ Para-Release™ Spray
Ease the release of paraffin blocks from embedding molds with Thermo Scientific™ Shandon™ Para-Release™ Spray.
Manufacturer:  Thermo Scientific™ 9990420
Catalog No. 9990420
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ease the release of paraffin blocks from embedding molds with Thermo Scientific™ Shandon™ Para-Release™ Spray.
  • Apply spray-releasing agent to molds before adding paraffin for clean release of block.
Specifications
Specifications
50mL