Learn More
Epredia™ Precision Cut Paraffin Available on GSA/VA Contract for Federal Government customers only.
Achieve thorough tissue infiltration and provide maximum support during microtomy with high-quality Precision Cut Paraffin. Also available in prepackaged kit for Epredia™ Excelsior™ AS and Revos™ Tissue Processors.
Manufacturer:  Epredia™ B1002490
 View more versions of this product
Catalog No. B1002490
Add to cart
Description
Description
Achieve thorough tissue infiltration and provide maximum support during microtomy with high-quality Epredia™ Paraffin.
  • Quality blend of paraffin, polymers and plastics (to varying degrees, based upon type)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
56° to 58°C
Embedding
Precision Cut
4 x 2.5 lb.