Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Shandon™ Polyethylene Dissecting Boards

Surgical cutting edges stay sharp on these polyethylene dissecting boards, available in white opaque or photographic blue surfaces.

Specifications

Material Polyethylene
Height (English) 0.75 in.
Height (Metric) 1.9cm
View More Specs

Products 1
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
B36115
View Documents
Thermo Scientific™
B36115
Blue Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Surgical cutting edges stay sharp on Thermo Scientific™ Shandon™ Polyethylene Dissecting Boards, available in white opaque or photographic blue surfaces.

Features:
  • Durable polyethylene board is 0.75 in. (1.9cm) thick and easy to clean.
  • White opaque surface provides an excellent background for cutting surgical or research specimens.
  • Photographic blue surface allows specimens to be photographed directly on the board.
Ordering information:
  • Large board measures 16 x 23 x 0.75 in. (40.6 x 58.4 x 1.9cm)
  • Small board measures 12 x 16 x 0.75 in. (30.48 x 40.64 x 1.905cm)
Specifications

Specifications

Polyethylene
0.75 in.
1.9cm
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit