Thermo Scientific™ Shandon™ Polysine Slides
Achieve superior cell and tissue adhesion with Thermo Scientific™ Shandon™ Polysine Slides.
Manufacturer: Thermo Scientific™ 6776215CS
 View more versions of this product
Catalog No. 67-762-15CS
  Check Availability
Add to Cart
    Specifications
    Color White
    Description White; 1440/Case
    Achieve superior cell and tissue adhesion with Thermo Scientific™ Shandon™ Polysine Slides.
    • Polysine places an advanced permanent adhesive on the microscope slide
    • Built-in chemicals electrostatically and chemically attract tissues and cells
  • Description & Specifications
Related Products
Product Content Feedback