Thermo Scientific™ Shandon™ Portable Waste Container
Built for heavy usage, the Shandon™ Portable Waste Container features durable, heavy-gauge stainless steel construction and includes locking cover and stand.
Manufacturer: Thermo Scientific™ 368
 View more versions of this product
Catalog No. 368
  Check Availability
Add to Cart
    Specifications
    Description Portable Waste Container
    Built for heavy usage, Thermo Scientific™ Shandon™ Portable Waste Container features durable, heavy-gauge stainless steel construction and includes locking cover and stand. Outisde dimensions measure 12 x 12.5 in. (30.5 x 31.8cm).
    • Durable, for heavy usage
    • 4-gallon capacity pail lifts off of stand
    • Stand supported by four rubber casters
    • Dimensions (O.D. x H): 12 x 12.5in. (30.5 x 31.8cm)
    Includes:Locking cover and stand (368 only)
  • Description & Specifications
Related Products
Product Content Feedback