Learn More
Thermo Scientific™ Shandon™ Rib Shears
Cut with Thermo Scientific™ Shandon™ Rib Shears, featuring curved blades and a single spring, for quality and precision.
Manufacturer:  Thermo Scientific™ 50219
 View more versions of this product
Catalog No. 50219
  Check Availability
Add to cart
Specifications
Description
For quality and precision, cut with Thermo Scientific™ Shandon™ Rib Shears, featuring curved blades and a single spring.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Guaranteed to be free from defects in material and workmanship
Description & Specifications
Specifications
Curved
9 in.
Locked, Separates at Pivot
Standard
22.9cm
Locked, Separates at Pivot; 22.9cm