Thermo Scientific™ Shandon™ Rochester-Pean's Hemostatic Forceps
Choose 6.25 in. (15.9cm) Thermo Scientific™ Shandon™ Rochester-Pean's Hemostatic Forceps with curved or straight arms for quality and precision.
Manufacturer: Thermo Scientific™ 5178
 View more versions of this product
Catalog No. 51-78
  Check Availability
Add to Cart
    Specifications
    Grade Standard
    Length (English) 6.25 in.
    Length (Metric) 15.9cm
    Shape Curved
    Type Rochester-Pean
    Choose 6.25 in. (15.9cm) Thermo Scientific™ Shandon™ Rochester-Pean's Hemostatic Forceps with curved or straight arms for quality and precision. Forceps are available in premium and standard grades.
    • Properly hardened, tempered and passivated
    • Electronically polished and cleaned for a high-quality finish
    • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
    • Guaranteed to be free from defects in material and workmanship
  • Description & Specifications
Related Products
Product Content Feedback