Learn More
Thermo Scientific™ Shandon™ Rochester-Pean's Hemostatic Forceps
Choose 6.25 in. (15.9cm) Thermo Scientific™ Shandon™ Rochester-Pean's Hemostatic Forceps with curved or straight arms for quality and precision.
Manufacturer:  Thermo Scientific™ 75152
 View more versions of this product
Catalog No. 75152
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Choose 6.25 in. (15.9cm) Thermo Scientific™ Shandon™ Rochester-Pean's Hemostatic Forceps with curved or straight arms for quality and precision. Forceps are available in premium and standard grades.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Guaranteed to be free from defects in material and workmanship
Specifications
Specifications
Straight
6.25 in.
Rochester-Pean Hemostatic Forceps
Premium
15.9cm