Thermo Scientific™ Shandon™ Russian Thumb Forceps
Properly hardened, tempered and passivated, choose these Russian Thumb Forceps with circular serrated tips for quality and precision.
Manufacturer: Thermo Scientific™ 6551
 View more versions of this product
Catalog No. 6551
  Check Availability
Add to Cart
    Specifications
    Grade Standard
    Length (English) 6 in.
    Length (Metric) 15.2cm
    Type Russian
    Properly hardened, tempered and passivated, choose Thermo Scientific™ Shandon™ Russian Thumb Forceps with circular serrated tips for quality and precision.
    Features:
    • Electronically polished and cleaned for a high-quality finish
    • Patterns, dimensions and finish held to exacting standards
    • Guaranteed to be free from defects in material and workmanship
    • Available in three lengths
    • Come in premium or standard grades
  • Description & Specifications
Related Products
Product Content Feedback