Learn More
Thermo Scientific™ Shandon™ Straight Blunt/Blunt Dissecting Scissors
Dissect using Thermo Scientific™ Shandon™ Straight Blunt/Blunt Dissecting Scissors, available in premium and standard grades.
Manufacturer:  Thermo Scientific™ 28203
 View more versions of this product
Catalog No. 28203
Add to cart
Description
Description
Dissect using Thermo Scientific™ Shandon™ Straight Blunt/Blunt Dissecting Scissors, available in premium and standard grades.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Guaranteed to be free from defects in material and workmanship
Specifications
Specifications
Straight
Standard
16.5cm
Blunt/Blunt
6.5 in.
Straight Blunt/Blunt Dissecting Scissors