Thermo Scientific™ Shandon™ Tissue-Processing/Embedding Cassettes II
Secure specimens during processing with Thermo Scientific™ Shandon™ Tissue Processing/Embedding Cassettes II.
Manufacturer: Thermo Scientific™ 1001054
 View more versions of this product
Catalog No. 10-010-54
  Check Availability
Add to Cart
    Specifications
    Pore Style Small Slots
    Material Acetal polymer
    Writing Face Angle 45°
    Color Gray
    Secure specimens during processing with Thermo Scientific™ Shandon™ Tissue Processing/Embedding Cassettes II, featuring a locking lid attached by two hinges that open from either the front or back.
    • Acetal polymer construction
    • Small slot-style pores
    • Cassette writing face is at a 45° angle
    • Box of 500 cassettes with attached lids
  • Description & Specifications
Related Products
Product Content Feedback