Learn More
Thermo Scientific™ Shandon™ Tissue-Processing/Embedding Cassettes II
Secure specimens during processing with Thermo Scientific™ Shandon™ Tissue Processing/Embedding Cassettes II.
Manufacturer:  Thermo Scientific™ 1001059
 View more versions of this product
Catalog No. 10-010-59
  Check Availability
Add to cart
Specifications
Description
Secure specimens during processing with Thermo Scientific™ Shandon™ Tissue Processing/Embedding Cassettes II, featuring a locking lid attached by two hinges that open from either the front or back.
  • Acetal polymer construction
  • Small slot-style pores
  • Cassette writing face is at a 45° angle
  • Box of 500 cassettes with attached lids
Description & Specifications
Specifications
Small Slots
45°
Green
Acetal polymer