Learn More
Thermo Scientific™ Shandon™ Tissue-Processing/Embedding Cassettes II
Secure specimens during processing with Thermo Scientific™ Shandon™ Tissue Processing/Embedding Cassettes II.
Specifications
Material Acetal polymer
Pore Style Small Slots
Writing Face Angle 45°
View More Specs
Products 10
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
10-010-52
View Documents
Thermo Scientific™
1001052
Orange This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
10-010-53
View Documents
Thermo Scientific™
1001053
Lilac This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
10-010-51
View Documents
Thermo Scientific™
1001051
Aqua This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
10-010-55
View Documents
Thermo Scientific™
1001055
Tan This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
10-010-56
View Documents
Thermo Scientific™
1001056
Peach This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
10-010-57
View Documents
Thermo Scientific™
1001057
Blue This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
10-010-58
View Documents
Thermo Scientific™
1001058
Yellow This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
10-010-59
View Documents
Thermo Scientific™
1001059
Green This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
10-010-60
View Documents
Thermo Scientific™
1001060
Pink This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
10-010-61
View Documents
Thermo Scientific™
1001061
White This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Secure specimens during processing with Thermo Scientific™ Shandon™ Tissue Processing/Embedding Cassettes II, featuring a locking lid attached by two hinges that open from either the front or back.
  • Acetal polymer construction
  • Small slot-style pores
  • Cassette writing face is at a 45° angle
  • Box of 500 cassettes with attached lids
Specifications
Specifications
Acetal polymer
Small Slots
45°