Learn More
Thermo Scientific™ Shandon™ Tissue Thumb Forceps
Choose Thermo Scientific™ Shandon™ Tissue Thumb Forceps with 1 x 2 teeth for quality and precision.
Manufacturer:  Thermo Scientific™ 6453
 View more versions of this product
Catalog No. 64-53
  Check Availability
Add to cart
Specifications
Description
Choose Thermo Scientific™ Shandon™ Tissue Thumb Forceps with 1 x 2 teeth for quality and precision. Forceps are available in premium and standard grades and come in four lengths.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Guaranteed to be free from defects in material and workmanship
  • Available in four lengths
Description & Specifications
Specifications
Standard
14cm
Tissue Thumb Forceps
5.5 in.
1 x 2