Learn More
Thermo Scientific™ Shandon™ Tissue Thumb Forceps
Choose Thermo Scientific™ Shandon™ Tissue Thumb Forceps with 1 x 2 teeth for quality and precision.
Specifications
Type Tissue Thumb Forceps
Teeth Count 1 x 2
View More Specs
Products 2
Catalog Number Mfr. No. Grade Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Grade Length (Metric) Price Quantity & Availability  
64-53
View Documents
Thermo Scientific™
6453
Standard 14cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
64-54
View Documents
Thermo Scientific™
6454
Standard 15.2cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Choose Thermo Scientific™ Shandon™ Tissue Thumb Forceps with 1 x 2 teeth for quality and precision. Forceps are available in premium and standard grades and come in four lengths.
Features:
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
  • Guaranteed to be free from defects in material and workmanship
Specifications
Specifications
Tissue Thumb Forceps
1 x 2