Learn More
Thermo Scientific™ Shandon™ Tissue Thumb Forceps
Choose Thermo Scientific™ Shandon™ Tissue Thumb Forceps with 2 x 3 Teeth for quality and precision.
Manufacturer:  Thermo Scientific™ 76603
 View more versions of this product
Catalog No. 76-603
  Check Availability
Add to cart
Specifications
Description
Choose Thermo Scientific™ Shandon™ Tissue Thumb Forceps with 2 x 3 Teeth for quality and precision. Forceps are available four lengths and come in premium and standard grades.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Guaranteed to be free from defects in material and workmanship
Description & Specifications
Specifications
Premium
15.2cm
Tissue Thumb Forceps
6 in.
2 x 3