Thermo Scientific™ Shandon™ Tissue Thumb Forceps with 3 x 4 Teeth
Choose Thermo Scientific™ Shandon™ Tissue Thumb Forceps with 3 x 4 Teeth for quality and precision.
Manufacturer: Thermo Scientific™ 76652
 View more versions of this product
Catalog No. 76-652
  Check Availability
Add to Cart
    Specifications
    Grade Premium
    Length (English) 5.5 in.
    Length (Metric) 14cm
    Teeth Count 3 x 4
    Type Thumb
    Choose Thermo Scientific™ Shandon™ Tissue Thumb Forceps with 3 x 4 Teeth for quality and precision. Forceps are available in four lengths and come in premium and standard grades.
    • Properly hardened, tempered and passivated
    • Electronically polished and cleaned for a high-quality finish
    • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
    • Guaranteed to be free from defects in material and workmanship
  • Description & Specifications
Related Products
Product Content Feedback