Thermo Scientific™ Shandon™ Tube Occluding Clamp Hemostatic Forceps
For quality and precision, choose Thermo Scientific™ Shandon™ Tube Occluding Clamp Hemostatic Forceps with straight arms and smooth jaws.
Manufacturer: Thermo Scientific™ 3120013
 View more versions of this product
Catalog No. 3120013
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Grade Standard
    Length (English) 4.5 in.
    Length (Metric) 11.4cm
    Shape Straight
    For quality and precision, choose Thermo Scientific™ Shandon™ Tube Occluding Clamp Hemostatic Forceps with straight arms and smooth jaws. Available in two lengths and in premium or standard grades.
    • Straight; smooth jaws
    • Properly hardened, tempered and passivated
    • Electronically polished and cleaned for a high-quality finish
    • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
    • Guaranteed to be free from defects in material and workmanship
  • Description & Specifications
Related Products
Product Content Feedback