Learn More
Thermo Scientific™ Shandon™ Wright-Giemsa Stain Kit
Stain using eosinates of polychrome methylene blue, which yield rich color, with the Thermo Scientific™ Shandon™ Wright-Giemsa Stain Kit.
Manufacturer:  Thermo Scientific™ 9990715
 View more versions of this product
Catalog No. 9990715
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Stain using eosinates of polychrome methylene blue, which yield rich color, with the Thermo Scientific™ Shandon™ Wright-Giemsa Stain Kit.
  • Produces an intense differential stain for blood smears, bone marrows, mast cells and FNA's
  • Demonstrates micro-organisms such as Microfilaria, Babesia, Haemobartonella, Giardia, Pneumocystis and Helicobacter
Specifications
Specifications
4L
Wright-Giemsa Solution