Thermo Scientific™ Shandon™ Zinc Formal-Fixx™ Available on GSA/VA Contract for Federal Government customers only.
Acquire amazing results with Thermo Scientific™ Shandon™ Zinc Formal-Fixx™, which is excellent for immunochemistry.
Manufacturer: Thermo Scientific™ 9990245
 View more versions of this product
Catalog No. 9990245
  Check Availability
Add to Cart
    Specifications
    Product Type Concentrated
    Quantity 2 x 1L,2 x 1L
    Type Concentrated
    Acquire amazing results with Thermo Scientific™ Shandon™ Zinc Formal-Fixx™, which is excellent for immunohistochemistry.
    • Enhances H&E's and special stains
    • Minimizes the need to enzyme digest
    • Reduces protein cross-linking
    • Produces well-defined nuclear detail
  • Description & Specifications
Related Products
Product Content Feedback