Learn More
Epredia™ Formal-Fixx&trade
Acquire amazing results with Epredia™ Formal-Fixx™, which is excellent for immunochemistry.
Supplier:  Epredia™ 6764250
 View more versions of this product
Catalog No. 6764250
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Enhances H&E's and special stains
  • Minimizes the need to enzyme digest
  • Reduces protein cross-linking
  • Produces well-defined nuclear detail
Specifications
Specifications
Fixative
19L