Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHANK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
$177.40 - $436.00
Specifications
Antigen | SHANK1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 -1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Description
SHANK1 Polyclonal specifically detects SHANK1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SHANK1 | |
Western Blot 1:500 -1:1000 | |
Polyclonal | |
Rabbit | |
Neuroscience | |
PBS with 0.02% Sodium Azide | |
50944 | |
A synthetic peptide made to an N terminal portion of human SHANK1 protein (between residues 110-160) [UniProt Q9Y566] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
SH3 and multiple ankyrin repeat domains 1, SH3 and multiple ankyrin repeat domains protein 1, Shank1, Somatostatin receptor-interacting protein, SSTR-interacting protein, SSTRIPSPANK-1, synamon | |
SHANK1 | |
IgG | |
Affinity Purified | |
Reacts with residues [ERLLREYPQSFEKGV] of the N terminus of the rat Shank1a protein. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title