Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHANK3 Mouse anti-Human, Mouse, Rat, FITC, Clone: S367-51, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242189F

Catalog No. NBP242189F


Add to cart

Description

Description

SHANK3 Monoclonal antibody specifically detects SHANK3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

SHANK3
S367-51
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
SHANK3
Fusion protein amino acids 538-626 (SH3 domain) of rat SHANK3. Mouse: 100% identity (89/89 amino acids identical). Human: 97% identity (87/89 amino acids identical). ~70% identity with SHANK1 and SHANK2.
Protein G purified
Store at 4C in the dark.
Monoclonal
Approx 70% identity with SHANK1 and SHANK2.Detects approx 190kDa. Cross-reacts with SHANK1. Does not cross-react with SHANK2.
Immunohistochemistry, Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
DEL22q13.3, KIAA1650proline rich synapse associated protein 2, Proline-rich synapse-associated protein 2, ProSAP2, PSAP2SH3 and multiple ankyrin repeat domains protein 3, SH3 and multiple ankyrin repeat domains 3, shank postsynaptic density protein, Shank3, SPANK-2
Mouse
IgG2a
0.1 ml
Primary
85358
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit